accessibility menu, dialog, popup

UserName

CAS RN 13734-40-2

(R)OHO(S)HNH3COOOCH3CH3CH3H3CCH3CH3

CAS RN 13734-40-2

IUPAC Name: 3-(tert-butoxy)-2-{[(tert-butoxy)carbonyl]amino}butanoic acid
Synonyms: 3-(tert-butoxy)-2-[(tert-butoxycarbonyl)amino]butanoic acid
Molecular Weight (g/mol): 275.35
Molecular Formula: C13H25NO5
InChi Key: LKRXXARJBFBMCE-UHFFFAOYNA-N
SMILES: CC(OC(C)(C)C)C(NC(=O)OC(C)(C)C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)