accessibility menu, dialog, popup

UserName

CAS RN 1373920-61-6

CH3FFFFOH

CAS RN 1373920-61-6

IUPAC Name: 4-fluoro-3-methyl-5-(trifluoromethyl)phenol
Synonyms: 4-fluoro-3-methyl-5-(trifluoromethyl)phenol
Molecular Weight (g/mol): 194.13
Molecular Formula: C8H6F4O
InChi Key: ZYLHFHFPFCTERO-UHFFFAOYSA-N
SMILES: CC1=CC(O)=CC(=C1F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results