accessibility menu, dialog, popup

UserName

CAS RN 1373921-12-0

H3COFOHOF

CAS RN 1373921-12-0

IUPAC Name: 2-(4-butoxy-2,6-difluorophenyl)acetic acid
Synonyms: (4-butoxy-2,6-difluorophenyl)acetic acid
Molecular Weight (g/mol): 244.24
Molecular Formula: C12H14F2O3
InChi Key: BOYCWRRKHQPVRH-UHFFFAOYSA-N
SMILES: CCCCOC1=CC(F)=C(CC(O)=O)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results