accessibility menu, dialog, popup

UserName

CAS RN 1373921-14-2

CH3OFFFF

CAS RN 1373921-14-2

IUPAC Name: 4-fluoro-3-methyl-5-(trifluoromethyl)benzaldehyde
Synonyms: 4-fluoro-3-methyl-5-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 206.14
Molecular Formula: C9H6F4O
InChi Key: FWHHVASQHPGQBB-UHFFFAOYSA-N
SMILES: CC1=C(F)C(=CC(C=O)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results