accessibility menu, dialog, popup

UserName

CAS RN 13754-41-1

NONH

CAS RN 13754-41-1

IUPAC Name: 4-benzylpiperazin-2-one
Synonyms: 4-benzylpiperazin-2-one
Molecular Weight (g/mol): 190.25
Molecular Formula: C11H14N2O
InChi Key: SBWVHKNCFZRBRJ-UHFFFAOYSA-N
SMILES: O=C1CN(CC2=CC=CC=C2)CCN1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results