accessibility menu, dialog, popup

UserName

CAS RN 13820-95-6

NNNClNNRhClCl

CAS RN 13820-95-6

IUPAC Name: rhodium(3+) pentaamine trichloride
Synonyms: pentamminerhodium chloride
Molecular Weight (g/mol): 294.41
Molecular Formula: Cl3H15N5Rh
InChi Key: ICJGGTVWZZBROS-UHFFFAOYSA-K
SMILES: N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Rh+3]
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)