accessibility menu, dialog, popup

UserName

CAS RN 1384424-80-9

H3CONHNOHNNFFF

CAS RN 1384424-80-9

IUPAC Name: 3-(7-methoxyquinolin-4-yl)-1-[6-(trifluoromethyl)pyridin-2-yl]urea
Synonyms: 3-(7-methoxyquinolin-4-yl)-1-[6-(trifluoromethyl)pyridin-2-yl]urea
Molecular Weight (g/mol): 362.31
Molecular Formula: C17H13F3N4O2
InChi Key: VQPBIJGXSXEOCU-UHFFFAOYSA-N
SMILES: COC1=CC=C2C(NC(=O)NC3=CC=CC(=N3)C(F)(F)F)=CC=NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results