accessibility menu, dialog, popup

UserName

H3COBrOOCH3

CAS RN 139102-34-4

IUPAC Name: methyl 4-bromo-2-methoxybenzoate
Synonyms: methyl 4-bromo-2-methoxybenzoate
Molecular Weight (g/mol): 245.07
Molecular Formula: C9H9BrO3
InChi Key: WPGAGRPPDYAZAD-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(OC)C=C(Br)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results