accessibility menu, dialog, popup

UserName

CAS RN 13918-92-8

FFSOOCl

CAS RN 13918-92-8

IUPAC Name: 2,4-difluorobenzene-1-sulfonyl chloride
Synonyms: 2,4-difluorobenzenesulfonyl chloride
Molecular Weight (g/mol): 212.59
Molecular Formula: C6H3ClF2O2S
InChi Key: FJSAJUXIHJIAMD-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(C=C1)S(Cl)(=O)=O
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)