CAS RN 139272-66-5
CAS RN 139272-66-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Aceclofenac Methyl Ester, TRC
CAS: 139272-66-5 Molecular Formula: C17 H15 Cl2 N O4 Molecular Weight (g/mol): 368.21 Synonym: Methyl [[[2-[(2,6-Dichlorophenyl)amino]phenyl]acetyl]oxy]acetate,Aceclofenac Imp. D (EP),Methyl Ester of Aceclofenac IUPAC Name: (2-methoxy-2-oxoethyl) 2-[2-(2,6-dichloroanilino)phenyl]acetate SMILES: COC(=O)COC(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl
2
Aceclofenac EP Impurity D, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.