CAS RN 139272-67-6
CAS RN 139272-67-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Aceclofenac Ethyl Ester, TRC
CAS: 139272-67-6 Molecular Formula: C18 H17 Cl2 N O4 Molecular Weight (g/mol): 382.24 Synonym: Ethyl [[[2-[(2,6-Dichlorophenyl)amino]phenyl]acetyl]oxy]acetate,Aceclofenac Imp. E (EP),Ethyl Ester of Aceclofenac IUPAC Name: (2-ethoxy-2-oxoethyl) 2-[2-(2,6-dichloroanilino)phenyl]acetate SMILES: CCOC(=O)COC(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl
2
Ethyl [[[2-[(2,6-Dichlorophenyl)amino]phenyl]acetyl]oxy]acetate (Ethyl Ester of Aceclofenac), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.