accessibility menu, dialog, popup

UserName

CAS RN 140-18-1

ClOO

CAS RN 140-18-1

IUPAC Name: benzyl 2-chloroacetate
Synonyms: benzyl 2-chloroacetate
Molecular Weight (g/mol): 184.62
Molecular Formula: C9H9ClO2
InChi Key: SOGXBRHOWDEKQB-UHFFFAOYSA-N
SMILES: ClCC(=O)OCC1=CC=CC=C1

 No Refinements