accessibility menu, dialog, popup

UserName

H3CCH3OONH2HCl

CAS RN 14019-62-6

IUPAC Name: propan-2-yl 2-aminoacetate hydrochloride
Synonyms: isopropyl 2-aminoacetate hydrochloride
Molecular Weight (g/mol): 153.61
Molecular Formula: C5H12ClNO2
InChi Key: UBKCIXXGQRZHRO-UHFFFAOYSA-N
SMILES: Cl.CC(C)OC(=O)CN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results