CAS RN 14086-88-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Acetamido-3-deoxy-D-glucose, TRC
CAS: 14086-88-5 Molecular Formula: C8 H15 N O6 Molecular Weight (g/mol): 221.21 IUPAC Name: N-[(3R,4S,5S,6R)-2,3,5-trihydroxy-6-(hydroxymethyl)oxan-4-yl]acetamide SMILES: CC(=O)N[C@@H]1[C@@H](O)C(O)O[C@H](CO)[C@H]1O