CAS RN 1414662-67-1
CAS RN 1414662-67-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
2'-Amino-5'-(4-carboxyphenyl)-[1,1':3',1' '-terphenyl]-4,4' '-dicarboxylic acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2'-Amino-5'-(4-carboxyphenyl)-[1,1':3',1' '-terphenyl]-4,4' '-dicarboxylic acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2'-Amino-5'-(4-carboxyphenyl)-[1,1':3',1' '-terphenyl]-4,4' '-dicarboxylic acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.