CAS RN 142784-25-6
CAS RN 142784-25-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(Deoxyguanosin-8-yl)-2-amino-1-methyl-6-phenylimidazo[4,5-Beta]pyridine, TRC
CAS: 142784-25-6 Molecular Formula: C23 H23 N9 O4 Molecular Weight (g/mol): 489.49 IUPAC Name: 9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-2-imino-8-[(1-methyl-6-phenylimidazo[4,5-b]pyridin-2-yl)amino]-3H-purin-6-one SMILES: Cn1c(Nc2nc3C(=O)NC(=N)Nc3n2[C@H]4C[C@H](O)[C@@H](CO)O4)nc5ncc(cc15)c6ccccc6