accessibility menu, dialog, popup

UserName

BOONH2

CAS RN 14335-29-6

IUPAC Name: diphenylboranyl 2-aminoacetate
Synonyms: diphenylboranyl 2-aminoacetate
Molecular Weight (g/mol): 239.08
Molecular Formula: C14H14BNO2
InChi Key: IVHLNYVPKWICDY-UHFFFAOYSA-N
SMILES: NCC(=O)OB(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results