accessibility menu, dialog, popup

UserName

CAS RN 14366-74-6

BrBrP

CAS RN 14366-74-6

IUPAC Name: {[4-(bromomethyl)phenyl]methyl}triphenylphosphanium bromide
Synonyms: {[4-(bromomethyl)phenyl]methyl}triphenylphosphanium bromide
Molecular Weight (g/mol): 526.25
Molecular Formula: C26H23Br2P
InChi Key: KUXBEOGHKWNPTG-UHFFFAOYSA-M
SMILES: [Br-].BrCC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results