CAS RN 1439-07-2
CAS RN 1439-07-2
IUPAC Name:
(2R,3R)-2,3-diphenyloxirane
Synonyms:
R-trans-stilbene oxide (R,R)-stilbene oxide
Molecular Weight (g/mol):
196.25
Molecular Formula:
C14H12O
InChi Key:
ARCJQKUWGAZPFX-ZIAGYGMSSA-N
SMILES:
O1[C@@H]([C@H]1C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
trans-Stilbene oxide, 99%, Thermo Scientific Chemicals
trans-Stilbene oxide, 99%, Thermo Scientific Chemicals
CAS: 1439-07-2 Molecular Formula: C14H12O Molecular Weight (g/mol): 196.25 MDL Number: MFCD00064311 InChI Key: ARCJQKUWGAZPFX-KWCCSABGSA-N Synonym: trans-stilbene oxide,3r-2,3-diphenyloxirane,2r-2alpha,3-diphenyloxirane PubChem CID: 5742860 IUPAC Name: (3R)-2,3-diphenyloxirane SMILES: C1=CC=C(C=C1)C2C(O2)C3=CC=CC=C3