accessibility menu, dialog, popup

UserName

OHBOHHN

CAS RN 144104-59-6

IUPAC Name: (1H-indol-5-yl)boronic acid
Synonyms: 1H-indol-5-ylboronic acid
Molecular Weight (g/mol): 160.97
Molecular Formula: C8H8BNO2
InChi Key: VHADYSUJZAPXOW-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C2NC=CC2=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)