accessibility menu, dialog, popup

UserName

CAS RN 1444-65-1

O

CAS RN 1444-65-1

IUPAC Name: 2-phenylcyclohexan-1-one
Synonyms: 2-phenylcyclohexan-1-one
Molecular Weight (g/mol): 174.24
Molecular Formula: C12H14O
InChi Key: DRLVMOAWNVOSPE-UHFFFAOYNA-N
SMILES: O=C1CCCCC1C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results