CAS RN 14504-60-0
CAS RN 14504-60-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5'-O-Methylthymidine, TRC
CAS: 14504-60-0 Molecular Formula: C11H16N2O5 Molecular Weight (g/mol): 256.26 IUPAC Name: 1-[(2R,4S,5R)-4-hydroxy-5-(methoxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione SMILES: COC[C@H]1O[C@H](C[C@@H]1O)N2C=C(C)C(=O)NC2=O