CAS RN 1451392-99-6
CAS RN 1451392-99-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2,6-Dichloro-4-fluorophenylboronic acid, TRC
CAS: 1451392-99-6 Molecular Formula: C6H4BO2FCl2 Molecular Weight (g/mol): 208.81 Synonym: B-(2,6-Dichloro-4-fluorophenyl)boronic acid(2,6-Dichloro-4-fluorophenyl)boronic acid SMILES: OB(O)c1c(Cl)cc(F)cc1Cl