accessibility menu, dialog, popup

UserName

CAS RN 14533-84-7

FFFFFOOFFF

CAS RN 14533-84-7

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 2,2,2-trifluoroacetate
Synonyms: 2,3,4,5,6-pentafluorophenyl 2,2,2-trifluoroacetate
Molecular Weight (g/mol): 280.07
Molecular Formula: C8F8O2
InChi Key: VCQURUZYYSOUHP-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C(F)(F)F)C(F)=C1F
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)