accessibility menu, dialog, popup

UserName

CH3(R)OHNSOHOO(R)(S)OH(S)(R)HO(S)CH3(R)CH3

CAS RN 14605-22-2

IUPAC Name: 2-[(4R)-4-[(1R,4S,5aS,7R,9aS,11aR)-4,7-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanamido]ethane-1-sulfonic acid
Synonyms: tauroursodeoxycholic acid
Molecular Weight (g/mol): 499.71
Molecular Formula: C26H45NO6S
InChi Key: BHTRKEVKTKCXOH-AYSJQVDDSA-N
SMILES: C[C@H](CCC(=O)NCCS(O)(=O)=O)[C@H]1CCC2C3[C@@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)C3CC[C@]12C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results