CAS RN 1464-44-4
CAS RN 1464-44-4
IUPAC Name:
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-phenoxyoxane-3,4,5-triol
Synonyms:
phenyl β-D-glucoside (α-(d))-phenylglucoside phenol glucoside (α-(D))-isomer phenylglucoside (α-(l))-isomer phenylglucoside phenylglucoside (α-(L))-isomer phenylglucoside (α-(d))-isomer phenylglucoside phenyl-β-glucoside aryl β-D-glucoside
Molecular Weight (g/mol):
256.25
Molecular Formula:
C12H16O6
InChi Key:
NEZJDVYDSZTRFS-RMPHRYRLSA-N
SMILES:
OC[C@H]1O[C@@H](OC2=CC=CC=C2)[C@H](O)[C@@H](O)[C@@H]1O
1
–
1
of
1
results
1
Phenyl beta-D-glucopyranoside, 98%
CAS: 1464-44-4 Molecular Formula: C12H16O6 Molecular Weight (g/mol): 256.25 MDL Number: MFCD03410292 InChI Key: NEZJDVYDSZTRFS-WLEIBRHLNA-N Synonym: phenyl beta-d-glucopyranoside,phenylglucoside,phenyl-beta-d-glucopyranoside,phenol glucoside,phenyl beta-d-glucoside,phenyl-beta-d-glucoside,.beta.-d-glucopyranoside, phenyl,unii-4w3pgi3766,aryl beta-d-glucoside,glucopyranoside, phenyl-, beta-d PubChem CID: 65080 ChEBI: CHEBI:28749 IUPAC Name: (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-phenoxyoxane-3,4,5-triol SMILES: OC[C@H]1O[C@@H](OC2=CC=CC=C2)[C@H](O)[C@@H](O)[C@@H]1O