accessibility menu, dialog, popup

UserName

CH3SOOOHH3CNOOCH

CAS RN 147202-94-6

IUPAC Name: 4-methylbenzene-1-sulfonic acid; prop-2-yn-1-yl 1-methyl-1,2,5,6-tetrahydropyridine-3-carboxylate
Synonyms: para-toluene sulfonate; prop-2-yn-1-yl 1-methyl-5,6-dihydro-2H-pyridine-3-carboxylate
Molecular Weight (g/mol): 351.42
Molecular Formula: C17H21NO5S
InChi Key: LYHWIOMAWPVTPI-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(O)(=O)=O.CN1CCC=C(C1)C(=O)OCC#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results