accessibility menu, dialog, popup

UserName

OHOO(E)

CAS RN 14755-02-3

IUPAC Name: (2E)-3-(3-hydroxyphenyl)prop-2-enoate
Synonyms: 3-hydroxycinnamate
Molecular Weight (g/mol): 163.15
Molecular Formula: C9H7O3
InChi Key: KKSDGJDHHZEWEP-SNAWJCMRSA-M
SMILES: OC1=CC=CC(\C=C\C([O-])=O)=C1
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)