CAS RN 1476-98-8
CAS RN 1476-98-8
IUPAC Name:
hydrogen {5-ethyl-1-azabicyclo[2.2.2]octan-2-yl}(6-methoxyquinolin-4-yl)methanol chloride
Synonyms:
dihydroquinidine
Molecular Weight (g/mol):
362.9
Molecular Formula:
C20H27ClN2O2
InChi Key:
MULXTQKDWYBJMO-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].CCC1CN2CCC1CC2C(O)C1=C2C=C(OC)C=CC2=NC=C1
1
–
2
of
2
results
1
Hydroquinidine Hydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Hydroquinidine Hydrochloride, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.