accessibility menu, dialog, popup

UserName

OOHH2NH2NHCl

CAS RN 1482-97-9

IUPAC Name: 2,3-diaminopropanoic acid hydrochloride
Synonyms: 2,3-diaminopropanoic acid hydrochloride
Molecular Weight (g/mol): 140.57
Molecular Formula: C3H9ClN2O2
InChi Key: SKWCZPYWFRTSDD-UHFFFAOYNA-N
SMILES: Cl.NCC(N)C(O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results