CAS RN 14920-87-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Thermo Scientific™ Methyl 2,6-dichlorobenzoate, 97%
CAS: 14920-87-7 Molecular Formula: C8H6Cl2O2 Molecular Weight (g/mol): 205.03 MDL Number: MFCD01313701 InChI Key: HEYGSGDIWJDORA-UHFFFAOYSA-N Synonym: 2,6-Dichlorobenzoic acid methyl ester; IUPAC Name: methyl 2,6-dichlorobenzoate SMILES: COC(=O)C1=C(Cl)C=CC=C1Cl