accessibility menu, dialog, popup

UserName

N(R)NO(S)(R)NNNH2HOHOBr

CAS RN 14985-44-5

IUPAC Name: 5-(6-amino-8-bromo-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Synonyms: 5-(6-amino-8-bromopurin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Molecular Weight (g/mol): 330.14
Molecular Formula: C10H12BrN5O3
InChi Key: NJBIVXMQFIQOGE-UHFFFAOYNA-N
SMILES: NC1=C2N=C(Br)N(C3CC(O)C(CO)O3)C2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results