accessibility menu, dialog, popup

UserName

CAS RN 15026-17-2

CH3H3CCH3OOOHO

CAS RN 15026-17-2

IUPAC Name: 4-(tert-butoxy)-4-oxobutanoic acid
Synonyms: 4-(tert-butoxy)-4-oxobutanoic acid
Molecular Weight (g/mol): 174.2
Molecular Formula: C8H14O4
InChi Key: PCOCFIOYWNCGBM-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)CCC(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results