CAS RN 15087-01-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Estrone b-D-Glucuronide Sodium Salt (>97% by HPLC), TRC
CAS: 15087-01-1 Molecular Formula: C24 H29 O8 . Na Molecular Weight (g/mol): 468.47 Synonym: Estrone-B-D-Glucuronide Sodium Salt IUPAC Name: sodium;(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[[(8R,9S,13S,14S)-13-methyl-17-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-yl]oxy]oxane-2-carboxylate SMILES: [Na+].C[C@]12CC[C@H]3[C@@H](CCc4cc(O[C@@H]5O[C@@H]([C@@H](O)[C@H](O)[C@H]5O)C(=O)[O-])ccc34)[C@@H]1CCC2=O