accessibility menu, dialog, popup

UserName

CAS RN 152529-79-8

CH3NOHNONHNSOOOH

CAS RN 152529-79-8

IUPAC Name: 4-(2,6-dioxo-1-propyl-2,3,6,7-tetrahydro-1H-purin-8-yl)benzene-1-sulfonic acid
Synonyms: 4-(2,6-dioxo-1-propyl-3,7-dihydropurin-8-yl)benzenesulfonic acid
Molecular Weight (g/mol): 350.35
Molecular Formula: C14H14N4O5S
InChi Key: UYDRRQPGDSIMNU-UHFFFAOYSA-N
SMILES: CCCN1C(=O)NC2=C(NC(=N2)C2=CC=C(C=C2)S(O)(=O)=O)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results