accessibility menu, dialog, popup

UserName

CAS RN 1526-17-6

OHFNOO

CAS RN 1526-17-6

IUPAC Name: 2-fluoro-6-nitrophenol
Synonyms: 2-fluoro-6-nitrophenol
Molecular Weight (g/mol): 157.1
Molecular Formula: C6H4FNO3
InChi Key: HIGRXCJEFUYRNW-UHFFFAOYSA-N
SMILES: OC1=C(F)C=CC=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results