CAS RN 153034-88-9
IUPAC Name:
2-chloro-4-iodo-3-methylpyridine
Synonyms:
2-chloro-4-iodo-3-methylpyridine
Molecular Weight (g/mol):
253.47
Molecular Formula:
C6H5ClIN
InChi Key:
WHCRABJNJRAJIP-UHFFFAOYSA-N
SMILES:
CC1=C(I)C=CN=C1Cl
1
–
1
of
1
results
1
2-Chloro-4-iodo-3-methylpyridine, 95%
CAS: 153034-88-9 Molecular Formula: C6H5ClIN Molecular Weight (g/mol): 253.467 MDL Number: MFCD03095293 InChI Key: WHCRABJNJRAJIP-UHFFFAOYSA-N Synonym: 2-chloro-4-iodo-3-picoline,2-chloro-3-methyl-4-iodopyridine,2-chloro-4-iodo-3-methyl-pyridine,pyridine, 2-chloro-4-iodo-3-methyl,2-chloro-4-iodo-3-methylpyridne,pubchem17059,2-chloranyl-4-iodanyl-3-methyl-pyridine,2-chloro-4-iodo-3-methylpyridine,2-chloro-4-iodo-3-methyl-pyridine; 2-chloro-4-iodo-3-picoline PubChem CID: 10083628 IUPAC Name: 2-chloro-4-iodo-3-methylpyridine SMILES: CC1=C(C=CN=C1Cl)I