CAS RN 1532-71-4
IUPAC Name:
1-bromoisoquinoline
Synonyms:
1-bromoisoquinoline
Molecular Weight (g/mol):
208.06
Molecular Formula:
C9H6BrN
InChi Key:
YWWZASFPWWPUBN-UHFFFAOYSA-N
SMILES:
BrC1=C2C=CC=CC2=CC=N1
1
–
1
of
1
results
1
1-Bromoisoquinoline, 95+%
CAS: 1532-71-4 MDL Number: MFCD00234478 InChI Key: YWWZASFPWWPUBN-UHFFFAOYSA-N Synonym: isoquinoline, 1-bromo,1-bromo-isoquinoline,bromoisoquinoline,zlchem 702,pubchem6211,acmc-1bqnj,1-bromoisoquinoline,abbypharma ap-12-10888 PubChem CID: 640963 IUPAC Name: 1-bromoisoquinoline SMILES: C1=CC=C2C(=C1)C=CN=C2Br