CAS RN 15325-51-6
IUPAC Name:
(Z,Z)-N'1,N'3-dihydroxybenzene-1,3-dicarboximidamide
Synonyms:
(Z,Z)-N'1,N'3-dihydroxybenzene-1,3-dicarboximidamide
Molecular Weight (g/mol):
194.19
Molecular Formula:
C8H10N4O2
InChi Key:
JVXOIYJLJWAUQP-UHFFFAOYSA-N
SMILES:
N\C(=N/O)C1=CC(=CC=C1)C(\N)=N\O
1
–
1
of
1
results
1
Isophthalamidoxime, 97%, Thermo Scientific™
CAS: 15325-51-6 Molecular Formula: C8H10N4O2 Molecular Weight (g/mol): 194.194 MDL Number: MFCD16659870 InChI Key: JVXOIYJLJWAUQP-UHFFFAOYSA-N Synonym: isophthalamidoxime,1,3-diamidoximobenzene,benzene-1,3-dicarboxamidoxime,e,z-n'1,n'3-dihydroxybenzene-1,3-dicarboximidamide PubChem CID: 73995304 IUPAC Name: 1-N',3-N'-dihydroxybenzene-1,3-dicarboximidamide SMILES: C1=CC(=CC(=C1)C(=NO)N)C(=NO)N