accessibility menu, dialog, popup

UserName

BOOCH3H3CCH3CH3OH3COCH3(E/Z)

CAS RN 153737-25-8

IUPAC Name: 2-(3,3-diethoxyprop-1-en-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(3,3-diethoxyprop-1-en-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 256.15
Molecular Formula: C13H25BO4
InChi Key: ZXGPKUGXWIEUJA-UHFFFAOYSA-N
SMILES: CCOC(OCC)C=CB1OC(C)(C)C(C)(C)O1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)