CAS RN 154212-61-0
CAS RN 154212-61-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
N-[[N-Methyl-N-[(2-isopropyl]-4-thiazolyl)methyl)amino]carbonyl-L-valine Carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-[[N-Methyl-N-[(2-isopropyl]-4-thiazolyl)methyl)amino]carbonyl-L-valine Carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(2S)-3-Methyl-2-[[methyl[[2-(1-methylethyl)thiazol-4-yl]methyl]carbamoyl]amino]butanoic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4
N-[[N-Methyl-N-[(2-isopropyl]-4-thiazolyl)methyl)amino]carbonyl-L-valine Carboxylic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.