accessibility menu, dialog, popup

UserName

CAS RN 1551-44-6

H3COO

CAS RN 1551-44-6

IUPAC Name: cyclohexyl butanoate
Synonyms: cyclohexyl butyrate
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: VZHUBBUZNIULNM-UHFFFAOYSA-N
SMILES: CCCC(=O)OC1CCCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results