accessibility menu, dialog, popup

UserName

CH3BrNHN

CAS RN 156454-43-2

IUPAC Name: 5-bromo-7-methyl-1H-indazole
Synonyms: 5-bromo-7-methyl-1H-indazole
Molecular Weight (g/mol): 211.06
Molecular Formula: C8H7BrN2
InChi Key: OUNQFZFHLJLFAR-UHFFFAOYSA-N
SMILES: CC1=C2NN=CC2=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results