accessibility menu, dialog, popup

UserName

OH(S)OO(R)HO

CAS RN 15667-21-7

IUPAC Name: (3R,4S)-3,4-dihydroxyoxolan-2-one
Synonyms: (3R,4S)-3,4-dihydroxyoxolan-2-one
Molecular Weight (g/mol): 118.09
Molecular Formula: C4H6O4
InChi Key: SGMJBNSHAZVGMC-STHAYSLISA-N
SMILES: O[C@H]1COC(=O)[C@@H]1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results