accessibility menu, dialog, popup

UserName

CAS RN 157-06-2

NH2(R)NH2NNH2OHO

CAS RN 157-06-2

IUPAC Name: (2R)-2-amino-5-[(diaminomethylidene)amino]pentanoic acid
Synonyms: (2R)-2-amino-5-[(diaminomethylidene)amino]pentanoic acid
Molecular Weight (g/mol): 174.2
Molecular Formula: C6H14N4O2
InChi Key: ODKSFYDXXFIFQN-SCSAIBSYSA-N
SMILES: N[C@H](CCCN=C(N)N)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)