accessibility menu, dialog, popup

UserName

CAS RN 158435-41-7

BrClO

CAS RN 158435-41-7

IUPAC Name: 4-bromo-2-chlorobenzaldehyde
Synonyms: 4-bromo-2-chlorobenzaldehyde
Molecular Weight (g/mol): 219.46
Molecular Formula: C7H4BrClO
InChi Key: DHGPLNJITGVCSG-UHFFFAOYSA-N
SMILES: ClC1=C(C=O)C=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results