accessibility menu, dialog, popup

UserName

CAS RN 159020-59-4

IUPAC Name: [3-fluoro-5-(trifluoromethyl)phenyl]boronic acid
Synonyms: 3-fluoro-5-(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol): 207.92
Molecular Formula: C7H5BF4O2
InChi Key: WEMCWZGCSRGJGW-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(F)=CC(=C1)C(F)(F)F
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)