accessibility menu, dialog, popup

UserName

CH3NF

CAS RN 15931-15-4

IUPAC Name: 3-fluoro-2-methylpyridine
Synonyms: 3-fluoro-2-methylpyridine
Molecular Weight (g/mol): 111.12
Molecular Formula: C6H6FN
InChi Key: IRIHSZDBTACXCT-UHFFFAOYSA-N
SMILES: CC1=C(F)C=CC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results