accessibility menu, dialog, popup

UserName

CAS RN 159689-88-0

FFFOBr

CAS RN 159689-88-0

IUPAC Name: 1-(bromomethyl)-3-(trifluoromethoxy)benzene
Synonyms: 1-(bromomethyl)-3-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 255.03
Molecular Formula: C8H6BrF3O
InChi Key: QSIVWRRHVXSDNE-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=CC(CBr)=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)