accessibility menu, dialog, popup

UserName

CAS RN 1603-41-4

CH3NH2N

CAS RN 1603-41-4

IUPAC Name: 5-methylpyridin-2-amine
Synonyms: 2-amino-5-methylpyridine
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: CMBSSVKZOPZBKW-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)N=C1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Boiling Point 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)